Learn More
Research Products International Corp Tween 80 (Polyoxyethylenesorbitan, Monooleate), 500 Milliliters
SDP
Catalog No. 50213613
Encompass_Preferred
Fatty acid compostion: Oleic acid approximately 70%
Balance primarily linoleic; palmitic and stearic acids.
Catalog No. 50-213-613 Supplier Research Products International Corp Supplier No. P203900.5
Fatty acid compostion: Oleic acid approximately 70%
Balance primarily linoleic; palmitic and stearic acids.