Learn More
Medchemexpress LLC HY-Y1891 50mL , Tween 80 CAS:9005-65-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY189150ML
Encompass_Preferred
HY-Y1891 50mL Tween 80 CAS: 9005-65-6 Tween 80 (Polysorbate 80), a surfactant, has been widely used as a solvent for pharmacological experiments. Tween 80 can also reduce bacterial attachment and inhibit biofilm formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8956