Learn More
Selleck Chemical LLC Tween 80
SDP
Catalog No. 5000002687
Encompass_Preferred
Tween 80 is a polysorbate surfactant used for selective protein extraction and the isolation of nuclei from mammalian cell lines Tween 80 has been used in drug vehicle solutions for specific injections in mice and rats It has been used for the determination of esterase activity in bacterial cells
Catalog No. 50-000-02687 Supplier Selleck Chemical LLC Supplier No. S6702100ML5
Tween 80 is a polysorbate surfactant used for selective protein extraction and the isolation of nuclei from mammalian cell lines Tween 80 has been used in drug vehicle solutions for specific injections in mice and rats It has been used for the determination of esterase activity in bacterial cells