Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polysorbate 80

Catalog No. ac278632500
Change view
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC278632500 250 mL
AC278630010 1 L
AC278630025 2.5 L
AC278630100 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC278632500 Supplier Thermo Scientific Chemicals Supplier No. 278632500
Only null left
Add to Cart

Polysorbate 80, CAS Number-9005-65-6, Polyoxyethylene(20)sorbitan monooleate, 1L, 2mg KOH/g max., Yellow to Amber, 1.0800g/mL, 5ppm max., >110 deg.C, 65 to 80mg KOH/g, Authentic, 15, 7703, MFCD00082107, Glass bottle, Viscous Liquid At 25 deg.C, Indoors +20 deg.C, 1.4710 to 1.4730, 45 to 55mg KOH/g | CAS: 9005-65-6 | (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6 | 604.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Synonym Polyoxyethylene(20)sorbitan monooleate

Specifications

Color Yellow to Amber
Content And Storage Indoors +20°C
Density 1.0800 g/mL
Quantity 250 mL
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
MDL Number MFCD00082107
Packaging Type Glass bottle
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
Molecular Weight (g/mol) 604.82
Solubility Solubility in water: 100g/L. Other solubilities: 100g/L isopropyl alcohol, 10g/L xylene, insoluble in propylene glycol and mineral oil
Form Viscous Liquid
Chemical Name or Material Polysorbate 80
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.