Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Tween™ 80 (Polyoxyethylene (20) Monooleate), OmniPur™, Calbiochem™,

Catalog No. 94954L Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Plastic Bottle
Plastic Receptacle in Fiberboard Box
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
94954L 4 L Plastic Receptacle in Fiberboard Box
94-901-L 1 L Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 94954L Supplier MilliporeSigma™ Supplier No. 94954L
Add to Cart
Add to Cart

Used for solubilization of membrane proteins, enzymology, analytical and diagnostic work, as well as chromatography and electrophoresis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-65-6
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC24H44O6
Molecular Weight (g/mol) 604.82
MDL Number MFCD00082107
InChI Key HDTIFOGXOGLRCB-KTKRTIGZNA-N
Synonym Polyoxyethylene Sorbitan Monooleate
PubChem CID 44324407
IUPAC Name 2-{2-[3,5-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl (9E)-octadec-9-enoate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Viscous liquid
Quantity 4 L
Packaging Plastic Receptacle in Fiberboard Box
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.