Learn More
Crescent Chemical Co Inc TWEEN 20 MOL BIO GRADE
SDP
Catalog No. 50248484
Encompass_Preferred
Tween 20 molecular biology grade DNase/RNase not detected. HLB 16.7. Size - 100ML Storage Conditions - +15 C TO +30 C Catalog Number - 39796.01 CAS 9005-64-5
Catalog No. 50-248-484 Supplier Crescent Chemical Co Inc Supplier No. SE39796.01
Tween 20 molecular biology grade DNase/RNase not detected. HLB 16.7. Size - 100ML Storage Conditions - +15 C TO +30 C Catalog Number - 39796.01 CAS 9005-64-5