Learn More
Crescent Chemical Co Inc TWEEN 20 PURE
SDP
Supplier:  Crescent Chemical Co Inc SE37470.01
Encompass_Preferred
Tween 20 research grade, Ph. Eur., USP HLB 16.7. Non-ionic surfactant that effectively suppresses unspecific reactions between antibodies, antigens and other molecules (1, 2). Also used as a solubilizer in membrane chemistry (3) and for density centrifugat
Catalog No. 50-248-266