Learn More
Crescent Chemical Co Inc TWEEN 80 PURE
SDP
Catalog No. 50248268
Encompass_Preferred
Tween 80 research grade, Ph. Eur., USP/NF HLB 15.0; tested for use in tissue culture. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 37475.01 CAS 9005-65-6
Catalog No. 50-248-268 Supplier Crescent Chemical Co Inc Supplier No. SE37475.01
Tween 80 research grade, Ph. Eur., USP/NF HLB 15.0; tested for use in tissue culture. Size - 500G Storage Conditions - +15 C TO +30 C Catalog Number - 37475.01 CAS 9005-65-6