Learn More
Crescent Chemical Co Inc TWEEN 80 PURE
SDP
Supplier:  Crescent Chemical Co Inc SE37475.02
Encompass_Preferred
Tween 80 research grade, Ph. Eur., USP/NF HLB 15.0; tested for use in tissue culture. Size - 5KG Storage Conditions - +15 C TO +30 C Catalog Number - 37475.02 CAS 9005-65-6
Catalog No. 50-248-269