Learn More
Ted Pella Inc TWEEZER
SDP
Catalog No. NC9304267
Encompass
Dumont High Precision Grade Tweezer, Fine points, Bent at 45degree, 105mm L, Points 0.1mm W x 0.06mm Thickness, Non-magnetic Stainless Steel, Style: 5/45, For electron microscopy, electronics, forensics, histology, and general laboratory
Catalog No. NC9304267 Supplier Ted Pella Inc Supplier No. 5724
Dumont High Precision Grade Tweezer, Fine points, Bent at 45degree, 105mm L, Points 0.1mm W x 0.06mm Thickness, Non-magnetic Stainless Steel, Style: 5/45, For electron microscopy, electronics, forensics, histology, and general laboratory