Learn More
Electron Microscopy Sciences EMS Gold Coated Tweezers Style 5TTHX, 118 mm OAL
SDP
Catalog No. 501927799
Encompass_Preferred
The EMS Gold Coated Tweezers are finely crafted, high precision tweezers made from anti-acidic, non-magnetic stainless steel, plated with 2 microns of pure 24-carat gold. Gold plating resists chemical corrosion, oxidation and is an excellent conductor of electricity. These tweezers are autoclavable and conducive for use when working with microelectronics, TEM and immunogold staining, electro-chemistry and nanotechnology. Reverse, Anti Capillary, Fine Point, High Precision.
OAL: 118 mm
Tip width: 0.10 mm
Tip thickness: 0.15 mm
Catalog No. 50-192-7799 Supplier Electron Microscopy Sciences Supplier No. 7875012
The EMS Gold Coated Tweezers are finely crafted, high precision tweezers made from anti-acidic, non-magnetic stainless steel, plated with 2 microns of pure 24-carat gold. Gold plating resists chemical corrosion, oxidation and is an excellent conductor of electricity. These tweezers are autoclavable and conducive for use when working with microelectronics, TEM and immunogold staining, electro-chemistry and nanotechnology. Reverse, Anti Capillary, Fine Point, High Precision.
OAL: 118 mm
Tip width: 0.10 mm
Tip thickness: 0.15 mm