Learn More
Electron Microscopy Sciences EMS Superalloy CX Biological Tweezers Style 3, 120 mm OAL
SDP
Catalog No. 501929477
Encompass_Preferred
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 120 mm
OAL: very fine sharp
Tip width: 0.04 mm
Catalog No. 50-192-9477 Supplier Electron Microscopy Sciences Supplier No. 783253SA
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 120 mm
OAL: very fine sharp
Tip width: 0.04 mm