Learn More
Electron Microscopy Sciences EMS Superalloy CX Biological Tweezers Style 5, 110 mm OAL
SDP
Catalog No. 501929479
Encompass_Preferred
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 110 mm
OAL: extra fine
Tip width: 0.02 mm
Catalog No. 50-192-9479 Supplier Electron Microscopy Sciences Supplier No. 783255SA
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 110 mm
OAL: extra fine
Tip width: 0.02 mm