Learn More
Electron Microscopy Sciences EMS Superalloy CX Biological Tweezers Style 7, 115 mm OAL
SDP
Catalog No. 501929480
Encompass_Preferred
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 115 mm
OAL: very fine curved
Tip width: 0.03 mm
Catalog No. 50-192-9480 Supplier Electron Microscopy Sciences Supplier No. 783257SA
EMS biological grade tweezers are made of a superalloy that contains nickel, chromium, and molybdenum. The Swiss made tweezers are anti-acid, anti-magnetic, and have serrated handles.
Tips: 115 mm
OAL: very fine curved
Tip width: 0.03 mm