Learn More
Electron Microscopy Sciences Style 2A, 1/EA
SDP
Catalog No. 502091057
Encompass_Preferred
These lightweight tweezers have Zirconia ceramic precision tips and aluminum handles. Surface resistively is 10E12 ohm. Contamination free, high temperature (1000°C).
Catalog No. 50-209-1057 Supplier Electron Microscopy Sciences Supplier No. 781272AT
These lightweight tweezers have Zirconia ceramic precision tips and aluminum handles. Surface resistively is 10E12 ohm. Contamination free, high temperature (1000°C).