Learn More
Electron Microscopy Sciences Style 71 MZ Ceramic 44mm,( Tips only)
SDP
Catalog No. 502091059
Encompass_Preferred
Ceramic replaceable tip tweezers with thin, fine tips. Stainless steel is anti-acid and anti-magnetic. OAL: 45 mm. Tips only, Tweezers sold separately.
Catalog No. 50-209-1059 Supplier Electron Microscopy Sciences Supplier No. 7812771T
Ceramic replaceable tip tweezers with thin, fine tips. Stainless steel is anti-acid and anti-magnetic. OAL: 45 mm. Tips only, Tweezers sold separately.