Learn More
Fine Science Tools Bulldog Serrefines, Straight, Stainless Steel, 35 mm
SDP
Catalog No. NC9208735
Encompass
Product: Bulldog Serrefine - Straight/35mm/ Shape: Straight/ Jaw Dimensions: 10.5 x 2.3mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 35mm/ Serrated: Yes/ Unit: Each
Catalog No. NC9208735 Supplier Fine Science Tools Supplier No. 1805035
Product: Bulldog Serrefine - Straight/35mm/ Shape: Straight/ Jaw Dimensions: 10.5 x 2.3mm/ Clamping Pressure: Strong/ Alloy: Stainless Steel/ Length: 35mm/ Serrated: Yes/ Unit: Each