Learn More
Electron Microscopy Sciences EMS Diamond Like Carbon Tweezer Set of 5 with Case
SDP
Supplier:  Electron Microscopy Sciences 7875214
Encompass_Preferred
Diamond like carbon (DLC) tweezers feature tips covered in a pure diamond film grown directly on the exposed metallic substrate via an innovative plasma-assisted deposition technique. It is not an adhesive-bonded diamond powder. The coating is comprised of carbon clusters which develop a crystalline structure and properties similar to a natural diamond. This procedure leaves the coating hydrogen free.
These tweezers perform well in demanding enviroments and working with hard samples or biological materials. Ideal for use in clean rooms. Tips stand up to repeated use. The body of the tweezer is non-magnetic, low carbon stainless steel. Set includes style 1, style 2A, style 3C, style 5, and style 7.
Catalog No. 50-192-7972