Learn More
Electron Microscopy Sciences EMS Diamond Like Carbon Tweezer Style 5B, 120 mm OAL
SDP
Catalog No. 501927968
Encompass_Preferred
Diamond like carbon (DLC) tweezers feature tips covered in a pure diamond film grown directly on the exposed metallic substrate via an innovative plasma-assisted deposition technique. It is not an adhesive-bonded diamond powder. The coating is comprised of carbon clusters which develop a crystalline structure and properties similar to a natural diamond. This procedure leaves the coating hydrogen free.
These tweezers perform well in demanding enviroments and working with hard samples or biological materials. Ideal for use in clean rooms. Tips stand up to repeated use. The body of the tweezer is non-magnetic, low carbon stainless steel. With Offset, Fine Tips.
OAL: 120 mm
Tip width: 0.12 mm
Tip thickness: 0.07 mm
Catalog No. 50-192-7968 Supplier Electron Microscopy Sciences Supplier No. 7875210
Diamond like carbon (DLC) tweezers feature tips covered in a pure diamond film grown directly on the exposed metallic substrate via an innovative plasma-assisted deposition technique. It is not an adhesive-bonded diamond powder. The coating is comprised of carbon clusters which develop a crystalline structure and properties similar to a natural diamond. This procedure leaves the coating hydrogen free.
These tweezers perform well in demanding enviroments and working with hard samples or biological materials. Ideal for use in clean rooms. Tips stand up to repeated use. The body of the tweezer is non-magnetic, low carbon stainless steel. With Offset, Fine Tips.
OAL: 120 mm
Tip width: 0.12 mm
Tip thickness: 0.07 mm