Learn More
World Precision Instrument Dumont Tweezers #3, 12cm, 0.4 x 0.3mm tips
SDP
Catalog No. 50822537
Encompass
Dumoxel Material Information
Resistant to staining, but slightly softer and less magnetic than stainless steel (Rockwell Hardness measurement of 36). Higher resistance to corrosion due to high content of molybdum and chromium. Very resistant to sulphuric acid, hydrochloric acid and other mineral and organic acids. Autoclavable. More likely to bend than to break. The maximum temperature this material can withstand is 350° C.
Dumostar Material Information
Highly resistant to metal fatigue, has great elasticity, and is durable (Rockwell Hardness measurement of 62). Resistant to mineral and organic acids, salt. Autoclavable. More likely to break than to bend. The maximum temperature this material can withstand is 550° C.
Catalog No. 50-822-537 Supplier World Precision Instrument Supplier No. 501208
 Restricted Item
Add to Cart
Add to Cart
Dumoxel Material Information
Resistant to staining, but slightly softer and less magnetic than stainless steel (Rockwell Hardness measurement of 36). Higher resistance to corrosion due to high content of molybdum and chromium. Very resistant to sulphuric acid, hydrochloric acid and other mineral and organic acids. Autoclavable. More likely to bend than to break. The maximum temperature this material can withstand is 350° C.
Dumostar Material Information
Highly resistant to metal fatigue, has great elasticity, and is durable (Rockwell Hardness measurement of 62). Resistant to mineral and organic acids, salt. Autoclavable. More likely to break than to bend. The maximum temperature this material can withstand is 550° C.