Learn More
Electron Microscopy Sciences Dumont Eyelash Tweezer Style 7SP, Inox 08, Matte Finish, 115 mm OAL
SDP
Catalog No. 501928080
Encompass_Preferred
Specially designed Dumont eyelash tweezers for the eyelash industry. Due to the demand and nature of the job to be done, we are directing those in the eyelash industry to this tweezer, which is perfect for the task.
Material: Inox 08
Finish: Matte
OAL: 115 mm
Tip width: 0.17
Tip thickness: 0.10
Catalog No. 50-192-8080 Supplier Electron Microscopy Sciences Supplier No. 727007SP
Specially designed Dumont eyelash tweezers for the eyelash industry. Due to the demand and nature of the job to be done, we are directing those in the eyelash industry to this tweezer, which is perfect for the task.
Material: Inox 08
Finish: Matte
OAL: 115 mm
Tip width: 0.17
Tip thickness: 0.10