Learn More
Electron Microscopy Sciences Dumont Brass Tweezers Style S5, Long, Brass
SDP
Catalog No. 501928648
Encompass_Preferred
These are popular tweezers for handling and manipulating delicate parts. Brass is composed of Cu, Pb, Sn, Ni and Zn. This alloy is 100% antimagnetic and is widely used for handling delicate components, especially in watch-making.
Catalog No. 50-192-8648 Supplier Electron Microscopy Sciences Supplier No. 5306S5PO
These are popular tweezers for handling and manipulating delicate parts. Brass is composed of Cu, Pb, Sn, Ni and Zn. This alloy is 100% antimagnetic and is widely used for handling delicate components, especially in watch-making.