Learn More
World Precision Instrument Economy Tweezers #2, 11cm, 0.4x0.55mm Tips
SDP
Catalog No. NC1608856
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. NC1608856 Supplier World Precision Instrument Supplier No. 501975
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.