Learn More
World Precision Instrument Economy Tweezers #5, 11cm, 0.4x0.45
SDP
Catalog No. NC1136358
Encompass
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.
Catalog No. NC1136358 Supplier World Precision Instrument Supplier No. 501979
Student fine tweezers are made of quality stainless steel. These tweezers are useful for general dissections where ultra fine tips are not necessary. Economically priced, they are affordable enough for everyone to have their own instruments.