Learn More
Electron Microscopy Sciences Dumont Replaceable Tip Tweezers Style WA1 Handle, All tips, 105 mm length
SDP
Catalog No. 501927482
Encompass_Preferred
These tweezer handles are anti-magnetic. Can be used with all tips with 4 screws. Length is 105 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tip width: 5.0 mm
Tip thickness: 5.0 mm
Length: 105 mm
Catalog No. 50-192-7482 Supplier Electron Microscopy Sciences Supplier No. 10004WA1SM
These tweezer handles are anti-magnetic. Can be used with all tips with 4 screws. Length is 105 mm. Resistant up to temperatures of 400 degree C (DIN 50 914) and suitable for autoclave sterilization at 270 degree C.
Tip width: 5.0 mm
Tip thickness: 5.0 mm
Length: 105 mm