Learn More
World Precision Instrument Jewelers #7 Forceps, 11cm, 0.07x0.01mm Tips, Titanium
SDP
Catalog No. NC1576619
Encompass
  • 11cm (4.33") Long
  • Curved
  • 0.07 x 0.01mm Fine Tips
  • Titanium
Catalog No. NC1576619 Supplier World Precision Instrument Supplier No. WP6570
  • 11cm (4.33") Long
  • Curved
  • 0.07 x 0.01mm Fine Tips
  • Titanium