Learn More
Electron Microscopy Sciences Dumont Style E10A ESD Replaceable Tweezer Tips, 0.6 mm Tip width, 0.5 mm Tip thickness, OAL
SDP
Catalog No. 501927500
Encompass_Preferred
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 0.6 mm
Tip thickness: 0.5 mm
Catalog No. 50-192-7500 Supplier Electron Microscopy Sciences Supplier No. TIP10AESD
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 0.6 mm
Tip thickness: 0.5 mm