Learn More
Electron Microscopy Sciences Dumont Style E15 ESD Replaceable Tweezer Tips, 5.35 mm Tip width, 0.3 mm Tip thickness, OAL
SDP
Catalog No. 501931620
Encompass_Preferred
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 5.35 mm
Tip thickness: 0.3 mm
Catalog No. 50-193-1620 Supplier Electron Microscopy Sciences Supplier No. TIP15ESD
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 5.35 mm
Tip thickness: 0.3 mm