Learn More
Electron Microscopy Sciences Dumont Style B159 Buxus Replaceable Tweezer Tips, 0.5 mm Tip width, 0.5 mm Tip thickness, OAL
SDP
Catalog No. 501927486
Encompass_Preferred
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 0.5 mm
Tip thickness: 0.5 mm
Catalog No. 50-192-7486 Supplier Electron Microscopy Sciences Supplier No. TIP159BUI
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 0.5 mm
Tip thickness: 0.5 mm