Learn More
Electron Microscopy Sciences Dumont Style E17B ESD Replaceable Tweezer Tips, 1.5 mm Tip width, 0.25 mm Tip thickness, OAL
SDP
Catalog No. 501931624
Encompass_Preferred
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 1.5 mm
Tip thickness: 0.25 mm
Catalog No. 50-193-1624 Supplier Electron Microscopy Sciences Supplier No. TIP17BESD
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 1.5 mm
Tip thickness: 0.25 mm