Learn More
Electron Microscopy Sciences Dumont Style E20k ESD Replaceable Tweezer Tips, 2.5 mm Tip width, 0.5 mm Tip thickness, 1.50 mm Tip C
SDP
Catalog No. 501927504
Encompass_Preferred
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 2.5 mm
Tip thickness: 0.5 mm
Tip C: 1.50 mm
Catalog No. 50-192-7504 Supplier Electron Microscopy Sciences Supplier No. TIP20ESD
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 2.5 mm
Tip thickness: 0.5 mm
Tip C: 1.50 mm