Learn More
Electron Microscopy Sciences Dumont Style E22 Black ESD Replaceable Tweezer Tips, 0.5 mm Tip width, 1 mm Tip thickness
SDP
Catalog No. 501927506
Encompass_Preferred
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 0.5 mm
Tip thickness: 1 mm
Tip C: 2.50 mm
Catalog No. 50-192-7506 Supplier Electron Microscopy Sciences Supplier No. TIP22ESD
Black ESD tips for WA1 tweezer handles.
Tips: Black ESD
Tip width: 0.5 mm
Tip thickness: 1 mm
Tip C: 2.50 mm