Learn More
Electron Microscopy Sciences Dumont Style B2A Buxus Replaceable Tweezer Tips, 1.5 mm Tip width, 0.2 mm Tip thickness, OAL
SDP
Catalog No. 501927492
Encompass_Preferred
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 1.5 mm
Tip thickness: 0.2 mm
Catalog No. 50-192-7492 Supplier Electron Microscopy Sciences Supplier No. TIP2ABUI
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 1.5 mm
Tip thickness: 0.2 mm