Learn More
Electron Microscopy Sciences Dumont Style BFSP Buxus Replaceable Tweezer Tips, 2 mm Tip width, 0.2 mm Tip thickness, OAL
SDP
Catalog No. 501927490
Encompass_Preferred
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 2 mm
Tip thickness: 0.2 mm
Catalog No. 50-192-7490 Supplier Electron Microscopy Sciences Supplier No. TIPFSPBUI
Boxwood (Buxus) tips for WA1 tweezer handles.
Tips: Boxwood (Buxus)
Tip width: 2 mm
Tip thickness: 0.2 mm