Learn More
Grainger Tweezer Set, Stainless Steel, 6 PIECES
Catalog No. NC2407465
Encompass_Preferred
This Tweezer Set is a set of 3 stainless steel tweezers designed for precision work. The set includes slant tip tweezers, pointed tip tweezers, and curved tip tweezers. The slant tip tweezers are ideal for grabbing small parts or manipulating wires. The pointed tip tweezers allow for precise grasping of tiny objects. The curved tip tweezers are useful for reaching into tight spaces. All three tweezers have a non-slip grip and stainless steel construction for durability. This tweezer set provides a variety of tip styles for detailed work.
Catalog No. NC2407465 Supplier Grainger Supplier No. 3LJZ3
This Tweezer Set is a set of 3 stainless steel tweezers designed for precision work. The set includes slant tip tweezers, pointed tip tweezers, and curved tip tweezers. The slant tip tweezers are ideal for grabbing small parts or manipulating wires. The pointed tip tweezers allow for precise grasping of tiny objects. The curved tip tweezers are useful for reaching into tight spaces. All three tweezers have a non-slip grip and stainless steel construction for durability. This tweezer set provides a variety of tip styles for detailed work.