Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style 00D, 120 mm OAL
SDP
Catalog No. 5023795
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Tips: serrated, thick and flat, medium sharp and strong for heavy duty work, serrated handle.
OAL: 120 mm
Catalog No. 50-237-95 Supplier Electron Microscopy Sciences Supplier No. 7813112
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Tips: serrated, thick and flat, medium sharp and strong for heavy duty work, serrated handle.
OAL: 120 mm