Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style 00D, 120 mm OAL
SDP
Supplier:  Electron Microscopy Sciences 7813101
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Tips: serrated, thick and flat, medium sharp and strong for heavy duty work, serrated handle.
OAL: 120 mm
Catalog No. 50-237-94