Learn More
Electron Microscopy Sciences Rubis Standard SA Steel Tweezers Style OC9, 90 mm OAL
SDP
Catalog No. 5023784
Encompass_Preferred
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Flat fine points.
OAL: 90 mm
Catalog No. 50-237-84 Supplier Electron Microscopy Sciences Supplier No. 7815201
With more than fifty years of experience, RUBIS tweezers represents authentic Swiss quality. The material used in these tweezers is SA steel, which is acid-proof, stainless steel, 100% antimagnetic and it is resistant to HF and nitric acid. Flat fine points.
OAL: 90 mm