Learn More
Electron Microscopy Sciences EMS Tweezers Style 249 with Replaceable Carbon PEEK Tips, 130 mm OAL
SDP
Catalog No. 501930841
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Strong tips made of carbon PEEK.
Material: Carbon PEEK
OAL: 130 mm
Catalog No. 50-193-0841 Supplier Electron Microscopy Sciences Supplier No. 7832525
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Strong tips made of carbon PEEK.
Material: Carbon PEEK
OAL: 130 mm