Learn More
Electron Microscopy Sciences EMS Tweezers Style 2A with Replaceable Carbon PEEK Tips, 125 mm OAL
SDP
Catalog No. 501930847
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Reverse action with flat round tips.
Material: Carbon PEEK
OAL: 125 mm
Catalog No. 50-193-0847 Supplier Electron Microscopy Sciences Supplier No. 7832531
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips and anti-magnetic stainless steel handles. Replaceable tip tweezers offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Reverse action with flat round tips.
Material: Carbon PEEK
OAL: 125 mm