Learn More
GROBET FILE CO OF AMERICA INC Swiss Pattern Tweezer, Peer-Vigor Brand, Pattern #3
Catalog No. 0887550
Encompass_Preferred
Excellent tweezers for handling all miniature and micro-miniature parts. Shanks are tapered with beveled edges; points have been honed and are very sharp. Overall length 4-3/4 inches (121 mm).
Catalog No. 08-875-50 Supplier GROBET FILE CO OF AMERICA INC Supplier No. 57.417
Excellent tweezers for handling all miniature and micro-miniature parts. Shanks are tapered with beveled edges; points have been honed and are very sharp. Overall length 4-3/4 inches (121 mm).