Learn More
Electron Microscopy Sciences Dumont Style 17B Delrin Replaceable Tweezer Tips, 1.5 mm Tip width, 0.25 mm Tip thickness, OAL
SDP
Catalog No. 501927522
Encompass_Preferred
Delrin tips for WA1 tweezer handles.
Tips: White Delrin
Tip width: 1.5 mm
Tip thickness: 0.25 mm
Catalog No. 50-192-7522 Supplier Electron Microscopy Sciences Supplier No. TIP17BDEL
Delrin tips for WA1 tweezer handles.
Tips: White Delrin
Tip width: 1.5 mm
Tip thickness: 0.25 mm