Learn More
Electron Microscopy Sciences Dumont Style 3 Grey STD Replaceable Tweezer Tips, 2.3 mm Tip width, Tip thickness, 35 mm OAL
SDP
Catalog No. 501927530
Encompass_Preferred
Replacement standard tips for WA1 tweezer handles.
Tips: Grey STD
Tip width: 2.3 mm
OAL: 35 mm
Catalog No. 50-192-7530 Supplier Electron Microscopy Sciences Supplier No. TIP3STD
Replacement standard tips for WA1 tweezer handles.
Tips: Grey STD
Tip width: 2.3 mm
OAL: 35 mm