Learn More
Electron Microscopy Sciences EMS Replacement Carbon Fiber Tips Style 00,
SDP
Catalog No. 501930615
Encompass_Preferred
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips, anti-magnetic stainless steel handles and offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Straight, strong, thick tips.
Material: Carbon Fiber
Catalog No. 50-193-0615 Supplier Electron Microscopy Sciences Supplier No. 7832545
Specially designed for the electronics industry, these tweezers feature durable and self-aligning replacement tips, anti-magnetic stainless steel handles and offer a significant cost savings. Excellent for handling sensitive electronic components, and ceramic and glass parts where non-metallic non-scratching is critical. 5 inches long and ESD safe. Straight, strong, thick tips.
Material: Carbon Fiber