Learn More
Electron Microscopy Sciences EMS Tweezers Tweezer Kit, Titanium Alloy
SDP
Catalog No. 501930580
Encompass_Preferred
The tweezers are the Swiss highest quality. Titanium alloy has a relatively low density (4.5g/cm3), good mechanical properties and high use temperature (1600°F/870°C). It is non magnetic, has good corrosion resistance, and useful where a very high strength-to weight ratio is required.
Material: Titanium Alloy
Catalog No. 50-193-0580 Supplier Electron Microscopy Sciences Supplier No. 7832502
The tweezers are the Swiss highest quality. Titanium alloy has a relatively low density (4.5g/cm3), good mechanical properties and high use temperature (1600°F/870°C). It is non magnetic, has good corrosion resistance, and useful where a very high strength-to weight ratio is required.
Material: Titanium Alloy