Learn More
Microsolv Technology Corporation Tweezers, Polypropylene, Blue. 1 EA.
Catalog No. 502269751
Encompass_Preferred
These lightweight, plastic Tweezers are handy for picking light weight objects that should not be touched by human hands to avoid contamination.Useful for Membrane Filters, Sparging Stones, Weighing Paper and many other items around a Laboratory.
This item is not returnable. View return policy
Catalog No. 50-226-9751 Supplier Microsolv Technology Corporation Supplier No. 4610001T
These lightweight, plastic Tweezers are handy for picking light weight objects that should not be touched by human hands to avoid contamination.Useful for Membrane Filters, Sparging Stones, Weighing Paper and many other items around a Laboratory.