Learn More
Ted Pella Inc TWEEZERS ANTI-CAPILLARY STYLE
SDP
Catalog No. NC9013079
Encompass
DUMONT Biology Anti-Capillary Tweezers,Style- 4ACN,Length-108mm,MTL- SS,Points Width x Thickness- 0.07 x 0.02mm,Qty-1-9,a electron microscopy tweezer
Catalog No. NC9013079 Supplier Ted Pella Inc Supplier No. 5104
DUMONT Biology Anti-Capillary Tweezers,Style- 4ACN,Length-108mm,MTL- SS,Points Width x Thickness- 0.07 x 0.02mm,Qty-1-9,a electron microscopy tweezer