Learn More
Electron Microscopy Sciences EMS Flat Carbon Fiber Soft Tip Tweezer Style 51, 110 mm OAL, 10 mm Tip width
SDP
Catalog No. 501927800
Encompass_Preferred
The EMS soft tip tweezers feature scratch resistant carbon fiber tips, specially designed to handle sensitive objects. The body is made from low carbon austenitic, non-magnetic stainless steel. The tips are corrosion resistant to moderately aggressive chemicals, salts and acids. Optimal working temperatures are up to 350 degree C (662 degree F). For short term use, maximum temperature should not exceed 925 degree C (1,697 degree F). Typical applications include handling of sensitive and electronic components, soft materials, micro-mechanical parts, glass, quartz, ceramics and silicon wafers. The tweezers body is made from non-magnetic stainless steel. ESD safe.
OAL: 110 mm
Tip width: 10 mm
Catalog No. 50-192-7800 Supplier Electron Microscopy Sciences Supplier No. 7875101
The EMS soft tip tweezers feature scratch resistant carbon fiber tips, specially designed to handle sensitive objects. The body is made from low carbon austenitic, non-magnetic stainless steel. The tips are corrosion resistant to moderately aggressive chemicals, salts and acids. Optimal working temperatures are up to 350 degree C (662 degree F). For short term use, maximum temperature should not exceed 925 degree C (1,697 degree F). Typical applications include handling of sensitive and electronic components, soft materials, micro-mechanical parts, glass, quartz, ceramics and silicon wafers. The tweezers body is made from non-magnetic stainless steel. ESD safe.
OAL: 110 mm
Tip width: 10 mm