Learn More
Ted Pella Inc PELCO Flat Tip | 127.sa | 107mm | NM-SS | 5.2 x 0.12mm
SDP
Catalog No. NC9007756
Encompass
The PELCO Pro Flat and Pad Tip Tweezers are high quality, precision Swiss made with smooth, scratch proof tips, useful for handling delicate and senitive parts. Can be used in laboratory, electronics, conservation, forensic, thin film research and many other applications. The smooth, flat tips enable gentle handling and minimum pressure on delicate parts. Eight different styles are available to provide the optimum tweezers for the application. Made of non-magnetic stainless steel.
Catalog No. NC9007756 Supplier Ted Pella Inc Supplier No. 5392NM
The PELCO Pro Flat and Pad Tip Tweezers are high quality, precision Swiss made with smooth, scratch proof tips, useful for handling delicate and senitive parts. Can be used in laboratory, electronics, conservation, forensic, thin film research and many other applications. The smooth, flat tips enable gentle handling and minimum pressure on delicate parts. Eight different styles are available to provide the optimum tweezers for the application. Made of non-magnetic stainless steel.