Learn More
Ted Pella Inc TWEEZERS, FLAT, ROUNDED TIPS
SDP
Catalog No. NC1994079
Encompass
AQUARIUS Tweezers, flat, rounded tipsStyle- 2a Length- 115mm Points Width x Thickness- 1.40 x 0.25mm
Catalog No. NC1994079 Supplier Ted Pella Inc Supplier No. 5818
AQUARIUS Tweezers, flat, rounded tipsStyle- 2a Length- 115mm Points Width x Thickness- 1.40 x 0.25mm